
Ambroxol Hydrochloride

Synonym(s):
Ambroxol hydrochloride, 2-Amino-3,5-dibromo-N-(trans-4-hydroxycyclohexyl)benzylamine
Empirical Formula (Hill Notation):
C13H18Br2N2O · HCl
CAS Number:
Molecular Weight:
414.56
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3