Skip to Content
AZ
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aluminum/Magnesium
Products
Manufacturer
Physical Form
Diameter (mm)
Filter Products
Showing 1-20 of 20
Compare
Product Number
Description
Pricing
GF58534656
Aluminum/Magnesium,
tube, Al 97%/Mg 3%, outside diameter 0.75 mm, length 600 mm, wall thickness 0.125 mm
Expand
View Pricing
GF73075675
Aluminum/Magnesium,
foil, thickness 0.015 mm, Al97%/Mg 3%, size 50 × 50 mm, temper as rolled
Expand
View Pricing
GF24010085
Aluminum/Magnesium,
foil, Al 97%/Mg 3%, thickness 0.50 mm, size 100 × 100 mm, temper as rolled
Expand
View Pricing
GF20334169
Aluminum/Magnesium,
foil, Al 97%/Mg 3%, thickness 0.015 mm, size 100 × 100 mm, temper as rolled
Expand
View Pricing
GF83014613
Aluminum/Magnesium,
foil, thickness 0.50 mm, Al97%/Mg 3%, size 25 × 25 mm, temper as rolled
Expand
View Pricing
GF90939651
Aluminum/Magnesium,
foil, thickness 0.50 mm, Al97%/Mg 3%, diameter 10 mm, temper as rolled
Expand
View Pricing
GF10556423
Aluminum/Magnesium,
foil, Al 97%/Mg 3%, thickness 0.015 mm, size 200 × 200 mm, temper as rolled
Expand
View Pricing
GF43782600
Aluminum/Magnesium,
foil, Al 97%/Mg 3%, thickness 0.50 mm, 25 mm diameter, temper as rolled
Expand
View Pricing
GF77434465
Aluminum/Magnesium,
tube, outside diameter 0.75 mm, Al97%/Mg 3%, length 570 mm, wall thickness 0.125 mm
Expand
View Pricing
GF22877781
Aluminum/Magnesium,
foil, Al 97%/Mg 3%, thickness 0.015 mm, size 25 × 25 mm, temper as rolled
Expand
View Pricing
GF90800670
Aluminum/Magnesium,
foil, thickness 0.50 mm, Al97%/Mg 3%, diameter 4 mm, temper as rolled
Expand
View Pricing
GF02476391
Aluminum/Magnesium,
foil, Al 97%/Mg 3%, thickness 0.50 mm, size 50 × 50 mm, temper as rolled
Expand
View Pricing
GF17027883
Aluminum/Magnesium,
foil, Al 97%/Mg 3%, thickness 0.50 mm, 15 mm diameter, temper as rolled
Expand
View Pricing
GF05630914
Aluminum/Magnesium,
foil, Al 97%/Mg 3%, thickness 0.50 mm, 8 mm diameter, temper as rolled
Expand
View Pricing
GF96047310
Aluminum/Magnesium,
foil, thickness 0.015 mm, Al97%/Mg 3%, length 0.5 m, temper as rolled
Expand
View Pricing
GF29919493
Aluminum/Magnesium,
foil, Al 97%/Mg 3%, thickness 0.01 mm, length 0.5 m, coil width 220 mm
Expand
View Pricing
GF78541511
Aluminum/Magnesium,
foil, thickness 0.50 mm, Al97%/Mg 3%, diameter 6 mm, temper as rolled
Expand
View Pricing
GF57700706
Aluminum/Magnesium,
foil, Al 97%/Mg 3%, thickness 0.50 mm, size 150 × 150 mm, temper as rolled
Expand
View Pricing
GF85021613
Aluminum/Magnesium,
foil, thickness 0.50 mm, Al97%/Mg 3%, diameter 50 mm, temper as rolled
Expand
View Pricing
GF08865928
Aluminum/Magnesium,
foil, Al 97%/Mg 3%, thickness 0.0125 mm, length 0.5 m, temper as rolled
Expand
View Pricing