Skip to Content
AZ
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ace pressure tubes with plunger valves and thermowells
Products
Product Category
tubes (8)
Manufacturer
Binding Type
Capacity (mL)
Diameter (mm)
Length (cm)
OD (mm)
Color
Package Size
Sterility
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
Z400408
Ace pressure tubes with plunger valves and thermowells,
capacity ~15 mL
Expand
View Pricing
Z400467
Ace pressure tubes with plunger valves and thermowells,
volume ~60 mL
Expand
View Pricing
Z400424
Ace pressure tubes with plunger valves and thermowells,
capacity ~35 mL
Expand
View Pricing
Z400432
Ace pressure tubes with plunger valves and thermowells,
capacity ~100 mL
Expand
View Pricing
Z400459
Ace pressure tubes with plunger valves and thermowells,
capacity ~120 mL
Expand
View Pricing
Z400440
Ace pressure tubes with plunger valves and thermowells,
capacity ~38 mL
Expand
View Pricing
Z400416
Ace pressure tubes with plunger valves and thermowells,
tube capacity ~48 mL
Expand
View Pricing
Z400475
Ace pressure tubes with plunger valves and thermowells,
capacity ~185 mL
Expand
View Pricing