
Acamprosate impurity A

Synonym(s):
3-Amino-1-propanesulfonic acid, 3APS, Homotaurine, Tramiprosate
Linear Formula:
H2N(CH2)3SO3H
CAS Number:
Molecular Weight:
139.17
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing