
6-Hydroxypyridine-2-carboxylic acid

Synonym(s):
6-Hydroxypicolinic acid, 6-Hydroxypyridine-2-carboxylic acid
Empirical Formula (Hill Notation):
C6H5NO3
CAS Number:
Molecular Weight:
139.11
Products
Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing