5-Iodocytosine
Synonym(s):
4-Amino-2-hydroxy-5-iodopyrimidine
Empirical Formula (Hill Notation):
C4H4IN3O
CAS Number:
Molecular Weight:
237.00
Products
Brand
Biological Source
Formula Weight
Physical Form
Showing 1-1 of 1
Compare
Product Number
Description
Pricing