Skip to Content
Merck
4-tert-Octylphenol analytical standard

4-tert-Octylphenol

4-tert-Octylphenol analytical standard
Synonym(s):
4-(1,1,3,3-Tetramethylbutyl)phenol
Linear Formula:
(CH3)3CCH2C(CH3)2C6H4OH
CAS Number:
Molecular Weight:
206.32
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Gene Alias
Melting Point (°C)
Purity
Format
Application
Markush Class
Markush Group
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing