Skip to Content
Merck
4-Dimethylamino-4′-nitrostilbene ≥99.8% (T)

4-Dimethylamino-4′-nitrostilbene

4-Dimethylamino-4′-nitrostilbene ≥99.8% (T)
Synonym(s):
N,N-Dimethyl-4′-nitro-4-stilbenamine, DANS
Linear Formula:
(CH3)2NC6H4CH=CHC6H4NO2
CAS Number:
Molecular Weight:
268.31

Products

Formula Weight
Melting Point (°C)
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing