
4,4′,4′′-Tris(N-(naphthalen-2-yl)-N-phenyl-amino)-triphenylamine

Synonym(s):
4,4′,4′′-Tris[2-naphthyl(phenyl)amino]triphenylamine, 2-TNATA, 2TNATA
Empirical Formula (Hill Notation):
C66H48N4
CAS Number:
Molecular Weight:
897.11
Products
Brand
Formula Weight
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing