
2-Ketohexanoic acid sodium salt

Synonym(s):
α-Ketocaproic acid sodium salt, 2-Ketocaproic acid sodium salt
Linear Formula:
C6H9O3Na
CAS Number:
Molecular Weight:
152.12
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing