Skip to Content
Merck
1-Chloromethyl-4-fluoro-1,4-diazoniabicyclo[2.2.2]octane bis(tetrafluoroborate) >95% in F+ active

1-Chloromethyl-4-fluoro-1,4-diazoniabicyclo[2.2.2]octane bis(tetrafluoroborate)

1-Chloromethyl-4-fluoro-1,4-diazoniabicyclo[2.2.2]octane bis(tetrafluoroborate) >95% in F+ active
Synonym(s):
N-Chloromethyl-N′-fluorotriethylenediammonium bis(tetrafluoroborate), F-TEDA
Empirical Formula (Hill Notation):
C7H14B2ClF9N2
CAS Number:
Molecular Weight:
354.26

Products

Product Category
Brand
Sustainability
Feature
Formula Weight
Melting Point (°C)
Physical Form
Functional Group
Markush Class
Markush Group
Reaction Type
Reagent Type
Showing 1-1 of 1