Skip to Content
Merck
All Photos(1)

Key Documents

821982

TSKgel® Amide-80 (5 µm) HPLC Columns

L x I.D. 25 cm × 4.6 mm, carbamoyl phase

Synonym(s):

TSKgel® Amide-80 HR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711
eCl@ss:
32110501

product name

TSKgel® Amide-80 HPLC Column, phase carbamoyl, L × I.D. 25 cm × 4.6 mm, 5 μm particle size

material

stainless steel hardware

Agency

suitable for USP L68

description

Amide-80 HR

product line

TSKgel®

manufacturer/tradename

Tosoh 21982

parameter

0-100% organic solvent
10-80 °C temperature
150 bar max. pressure

technique(s)

HPLC: suitable

L × I.D.

25 cm × 4.6 mm

surface area

450 m2/g

matrix

silica particle platform

matrix active group

carbamoyl phase

particle size

5 μm

pore size

100 Å

operating pH

2.0-7.5

separation technique

hydrophilic interaction (HILIC)

Looking for similar products? Visit Product Comparison Guide

General description

TSKgel Amide-80 columns are now also available packed with 2 μm spherical silica particles, offering higher resolution and faster analysis with equivalent retention and selectivity as TSKgel Amide-80, 3 μm columns. This allows for smooth method transfer from a 3 μm to a 2 μm column. In addition, TSKgel Amide-80, 2 μm columns more highly retain hydrophilic compounds than competitive amide columns on the market.
Packed with 2, 3, 5, or 10 μm spherical silica particles that are covalently bonded with carbamoyl groups, TSKgel Amide-80 columns are used for the analysis of saccharides, glycans, oligosaccharides, peptides and polar compounds from natural products. The amide stationary phase provides a unique selectivity under regular normal phase conditions or in the hydrophilic interaction mode of chromatography.
TSKgel Amide-80 HR, 5 μm columns offer higher specifications for plate count compared to traditional TSKgel Amide-80, 5 μm, 4.6 mm ID x 25 cm columns (N = > 18,000 vs. 8,000). This equates to more efficient chromatography.

Physical form

Shipped in 75% acetonitrile - 25% water.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service