Skip to Content
Merck
All Photos(1)

Key Documents

SRP3178

Sigma-Aldrich

TPO human

Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

MGDF, Megakaryocyte colony-stimulating factor, Thrombopoietin, c-MPL Ligand

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

≤1.0 ng/mL ED50

mol wt

18.6 kDa

packaging

pkg of 10 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... THPO(7066)

General description

Thrombopoietin (TPO) is a lineage specific growth factor, produced in the liver, kidney and skeletal muscle. It is an acidic glycoprotein. The gene encoding it is localized on human chromosome 3q27.1. Recombinant human TPO is a fully biologically active 174 amino acid polypeptide (18.6kDa), which contains the erythropoietin-like domain of the full length TPO protein.

Biochem/physiol Actions

Thrombopoietin (TPO) stimulates the proliferation and maturation of megakaryocytes, and promotes increased circulating levels of platelets in vivo. TPO signals through the cellular proto-oncogene (c-mpl) receptor and acts as an important regulator of circulating platelets. High levels of TPO have been observed in patients with aplastic anemia. Human and murine TPO exhibits cross-species reactivity.

Sequence

SPAPPACDLR VLSKLLRDSH VLHSRLSQCP EVHPLPTPVL LPAVDFSLGE WKTQMEETKA QDILGAVTLL LEGVMAARGQ LGPTCLSSLL GQLSGQVRLL LGALQSLLGT QLPPQGRTTA HKDPNAIFLS FQHLLRGKVR FLMLVGGSTL CVRRAPPTTA VPSRTSLVLT LNEL

Physical form

Lyophilized from 0.1% TFA.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a stabilizer (example 5% Trehalose) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pancreatic Cancer Cells Induce MicroRNA Deregulation in Platelets.
D?-az-Blancas, et al.
International Journal of Molecular Sciences, 23 (2022)
Jiaxin Zhao et al.
Arteriosclerosis, thrombosis, and vascular biology, 41(10), 2523-2537 (2021-08-06)
Objective: Roxadustat is a new medication for the treatment of renal anemia. EPO (erythropoietin)-the current treatment standard-has been reported to enhance platelet activation and production. However, to date, the effect of roxadustat on platelets is unclear. To address this deficiency
Yoshio Takei
Handbook of Hormones: Comparative Endocrinology for Basic and Clinical Research (2015)
Kuter, D.J., et al.
Cytokine Reference, 965-982 (2001)
The molecular mechanisms that control thrombopoiesis.
Kaushansky K
The Journal of Clinical Investigation, 115(12), 3339-3347 (2005)

Articles

Read article on hematopoietic cytokines and hematopoiesis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service