Skip to Content
Merck
All Photos(1)

Key Documents

SAE0012

Sigma-Aldrich

Hypo-Opticlear Human Sera

Synonym(s):

Human Serum

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.25

biological source

human

Quality Level

composition

total protein, ≥6.0 g/dL

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

This human serum is processed to reduce both T3 and T4 to zero values, making these sera suitable for use in the preparation of standards and controls.
Delipidization increases shelf life, improves appearance and eliminates interference from turbidity or the presence of lipids.
Contains low enzymes and low serum iron.
This stripped serum can withstand numerous freeze-thaws without affecting clarity.

Analysis Note

Endotoxin tested

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiangfeng Lv et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 128, 110236-110236 (2020-05-25)
Integrin αvβ3 was reported as positive regulators of tumorigenesis and highly expressed in cancer stem cells and kinds of cancers, thus, it is an appealing target for cancer treatment. Nanomedicine with targeting delivery ability has developed rapidly and shown its

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service