Skip to Content
Merck
All Photos(7)

Key Documents

HPA007272

Sigma-Aldrich

Anti-KRT7 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CK-7 antibody produced in rabbit, Anti-Cytokeratin-7 antibody produced in rabbit, Anti-K7 antibody produced in rabbit, Anti-Keratin, type II cytoskeletal 7 antibody produced in rabbit, Anti-Keratin-7 antibody produced in rabbit, Anti-Sarcolectin antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human, mouse, rat

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

RQEESEQIKTLNNKFASFIDKVRFLEQQNKLLETKWTLLQEQKSAKSSRLPDIFEAQIAGLRGQLEALQVDGGRLEAELRSMQDVVEDFKNKYEDEINRRTAAENEFVVLKKDVD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KRT7(3855)

General description

KRT7 (Keratin, type II cytoskeletal 7) gene encodes a type II cytokeratin with a molar mass of 55kDa. It is expressed during the differentiation of simple and stratified epithelial tissues. It is also found in the epithelia lining the cavities of internal organs such as stomach.

Immunogen

Keratin, type II cytoskeletal 7 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

KRT7 (Keratin, type II cytoskeletal 7) is associated with human tumor marker protein p150, the largest subunit of eukaryotic translation initiation factor 3. It serves as a predictive marker for survival in patients suffering from esophageal squamous cell carcinoma. In conjugation with specific growth factors or hormones, KRT7 restricts interferon-dependent interphase followed by the stimulation of DNA synthesis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70151

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Glass et al.
The Journal of cell biology, 101(6), 2366-2373 (1985-12-01)
Using mRNA from cultured human mesothelial cells, we constructed bacterial plasmids and lambda phage vectors that contained cDNA sequences specific for the keratins expressed in these cells. A cloned cDNA encoding keratin K7 (55 kD) was identified by positive hybrid
A Kaba et al.
Biochimie, 81(7), 709-715 (1999-09-24)
Interferons (IFNs) are major cytokines, responsible for down-regulating cell growth and for promoting cell differentiation. The sarcolectin (SCL) protein presented here blocks in the cells the established IFN-dependent interphase and stimulates DNA synthesis, probably in co-ordination with more specific growth
L Lin et al.
Journal of cellular biochemistry, 80(4), 483-490 (2001-02-13)
The human tumor marker protein p150 was identified as the largest subunit of eukaryotic translation initiation factor 3 (eIF3) (also known as p170/p180). Its expression level is not only upregulated in many transformed cell lines, but also in several human
Naohide Oue et al.
Annals of surgical oncology, 19(6), 1902-1910 (2011-12-29)
Patients diagnosed with stage II and III esophageal squamous cell carcinoma (ESCC) have variable prognosis. This group would benefit greatly from the discovery of prognostic markers that are capable of identifying individuals for whom adjuvant treatment would be advantageous. The
Roberta Ragazzini et al.
Developmental cell, 58(22), 2428-2446 (2023-09-01)
Thymus is necessary for lifelong immunological tolerance and immunity. It displays a distinctive epithelial complexity and undergoes age-dependent atrophy. Nonetheless, it also retains regenerative capacity, which, if harnessed appropriately, might permit rejuvenation of adaptive immunity. By characterizing cortical and medullary

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service