Skip to Content
Merck
All Photos(1)

Key Documents

B9647

Sigma-Aldrich

(E)-5-(2-Bromovinyl)-2′-deoxyuridine

Synonym(s):

BVdU

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H13BrN2O5
CAS Number:
Molecular Weight:
333.14
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

powder

Quality Level

storage temp.

2-8°C

SMILES string

O=C(C(/C=C/Br)=CN1[C@H]2C[C@H](O)[C@@H](CO)O2)NC1=O

InChI

1S/C11H13BrN2O5/c12-2-1-6-4-14(11(18)13-10(6)17)9-3-7(16)8(5-15)19-9/h1-2,4,7-9,15-16H,3,5H2,(H,13,17,18)/b2-1+/t7-,8+,9+/m0/s1

InChI key

ODZBBRURCPAEIQ-PIXDULNESA-N

Gene Information

human ... HV1S(3365)

Application

(E)-5-(2-Bromovinyl)-2′-deoxyuridine (BVdU) has been used:
  • as a substrate for thymidine kinase 1 to study its effects on cancer cells by a dual-promoter integrator approach
  • as a bromovinyl nucleoside analog to study its effects on varicella-zoster virus replication in a mouse model
  • as a nucleoside analog prodrug to study its effects on transduced cells

Biochem/physiol Actions

(E)-5-(2-Bromovinyl)-2′-deoxyuridine(BVdU) is a thymidine analog and a well-known inhibitor of Herpes simplex virus type 1 (HSV-1) and Varicella zoster virus (VZV) infections. It shows a minimal effect on HSV-2. BVdU shows therapeutic effects against herpetic keratitis, recurrent herpes labialis, and herpes zoster. BVdU undergoes phosphorylation at its 5’-diphosphate and further to 5’-triphosphate by the virus-encoded thymidine kinase, this results in BVdU acting as a competitive inhibitor to the viral DNA polymerase.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

(E)-5-(2-Bromovinyl)-2'-Deoxyuridine (BVDU)
Clercq E, et al.
Medical Research Engineering, 25(1), 1-20 (2005)
Jörg-Christian Heinrich et al.
Journal of cancer research and clinical oncology, 137(9), 1349-1361 (2011-08-13)
Several reports describe the importance of the chaperone HSP27 (HSPB1) in cancer progression, and the demand for drugs that modulate HSPB1-activity is increasing rapidly. We reported earlier that RP101 (Bromovinyldeoxyuridine, BVDU, Brivudine) improves the efficacy of chemotherapy in pancreatic cancer.
Tomoyuki Shiota et al.
Antiviral research, 91(2), 142-149 (2011-06-15)
Antiviral-resistant herpesvirus infection has become a great concern for immunocompromised patients. Herpes simplex virus type 1 (HSV-1) infections are treated with viral thymidine kinase (vTK)-associated drugs such as acyclovir (ACV), and most ACV-resistance (ACV(r)) is due to mutations in the
Leonardo D'Aiuto et al.
Schizophrenia bulletin, 41(1), 123-132 (2014-03-14)
Herpes simplex virus, type 1 (HSV-1) commonly produces lytic mucosal lesions. It invariably initiates latent infection in sensory ganglia enabling persistent, lifelong infection. Acute HSV-1 encephalitis is rare and definitive evidence of latent infection in the brain is lacking. However
Vineet Kumar et al.
Bioorganic & medicinal chemistry letters, 18(20), 5640-5642 (2008-09-18)
Nucleoside-based antiviral drugs have been synthesized using imidazolium-based ionic liquids as reaction medium. The ionic liquids were proved to be better solvents for all the nucleoside in terms of solubility and reaction medium as compared to conventional molecular solvents.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service