Skip to Content
Merck
All Photos(4)

Key Documents

228648

Sigma-Aldrich

Magnesium acetate tetrahydrate

ACS reagent, ≥98%

Synonym(s):

Acetic acid magnesium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3COO)2Mg · 4H2O
CAS Number:
Molecular Weight:
214.45
Beilstein:
3730605
EC Number:
MDL number:
UNSPSC Code:
12352106
eCl@ss:
39021909
PubChem Substance ID:
NACRES:
NB.24

grade

ACS reagent

Assay

≥98%
98.0-102.0% (ACS specification)

form

solid

impurities

≤0.001% N compounds
≤0.005% insolubles

mp

72-75 °C (lit.)

anion traces

chloride (Cl-): ≤0.001%
sulfate (SO42-): ≤0.005%

cation traces

Ba: ≤0.001%
Ca: ≤0.01%
Fe: ≤5 ppm
K: ≤0.005%
Mn: ≤0.001%
Na: ≤0.005%
Sr: ≤0.005%
heavy metals: ≤5 ppm (by ICP)

SMILES string

O.O.O.O.CC(=O)O[Mg]OC(C)=O

InChI

1S/2C2H4O2.Mg.4H2O/c2*1-2(3)4;;;;;/h2*1H3,(H,3,4);;4*1H2/q;;+2;;;;/p-2

InChI key

XKPKPGCRSHFTKM-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Magnesium acetate tetrahydrate can be used as:
  • A base to synthesize diarylbenzofuran carboxylic acids by reacting m-hydroxybenzoic acids and alkynes via Ru-catalyzed C-H alkenylation.
  • A magnesium source to synthesize porous magnesium carboxylate frameworks.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marie L Salva et al.
Small (Weinheim an der Bergstrasse, Germany), 16(49), e2005476-e2005476 (2020-11-18)
Nucleic acid hybridization reactions play an important role in many (bio)chemical fields, for example, for the development of portable point-of-care diagnostics, and often such applications require nucleic acid-based reaction systems that ideally run without enzymes under isothermal conditions. The use
Nobuyoshi Koga et al.
The journal of physical chemistry. B, 116(49), 14477-14486 (2012-11-20)
The kinetics and mechanism of the thermal dehydration of magnesium acetate tetrahydrate were investigated as a typical example of the glass formation process via the thermal decomposition of solids. Formation of an intermediate fluid phase was identified as the characteristic
The crystal structure of magnesium acetate-tetrahydrate Mg (CH3 COO)2. 4H2 O.
Shankar J, et al.
Proceedings of the Indian Academy of Sciences - Section A, 45(2), 117-117 (1957)
Hieu Bui et al.
Nano letters, 10(9), 3367-3372 (2010-08-05)
To fabricate quantum dot arrays with programmable periodicity, functionalized DNA origami nanotubes were developed. Selected DNA staple strands were biotin-labeled to form periodic binding sites for streptavidin-conjugated quantum dots. Successful formation of arrays with periods of 43 and 71 nm
Catarina M S S Neves et al.
Physical chemistry chemical physics : PCCP, 17(47), 32044-32052 (2015-11-18)
Due to scarce available experimental data, as well as due to the absence of predictive models, the influence of salts on the solubility of ionic liquids (ILs) in water is still poorly understood. To this end, this work addresses the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service