Skip to Content
Merck
All Photos(1)

Key Documents

Y0001108

Buprenorphine

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H41NO4
CAS Number:
Molecular Weight:
467.64
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

buprenorphine

manufacturer/tradename

EDQM

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada; psicótropo (Spain); Decreto Lei 15/93: Tabela IIC (Portugal)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

InChI

1S/C29H41NO4.ClH/c1-25(2,3)26(4,32)20-15-27-10-11-29(20,33-5)24-28(27)12-13-30(16-17-6-7-17)21(27)14-18-8-9-19(31)23(34-24)22(18)28;/h8-9,17,20-21,24,31-32H,6-7,10-16H2,1-5H3;1H/t20-,21-,24-,26+,27-,28+,29?;/m1./s1

InChI key

UAIXRPCCYXNJMQ-CHLZSUIASA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Buprenorphine EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 4 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takahiro Yoshizawa et al.
Experimental animals, 68(3), 277-283 (2019-02-15)
Locomotor activity is affected by a range of factors in addition to experimental treatment, including the breeding environment. Appropriate convalescence and acclimation are important for animal experiments, because environmental changes and physical burden can result from surgery, transportation, and cage
Damien A Devault et al.
Environmental science and pollution research international, 24(33), 25645-25655 (2016-01-16)
The consumption of drugs of abuse has been recently investigated in Martinique using the back-calculation approach, also called the "sewage epidemiology" method. Results demonstrated a very high consumption considering the international data. Wastewater treatment plants (WWTPs) are located just behind
Azadeh Esfandiary et al.
Cardiovascular research, 115(7), 1217-1227 (2019-03-10)
The role of uncoupling protein 2 (UCP2) in cardiac adaptation to pressure overload remains unclear. In a classical model of left ventricular pressure overload genetic deletion of UCP2 (UCP2-/-) protected against cardiac hypertrophy and failure. However, in UCP2-/- mice increased
Martin P Wegman et al.
The Lancet. Global health, 5(2), e198-e207 (2016-12-15)
Detention of people who use drugs into compulsory drug detention centres (CDDCs) is common throughout East and Southeast Asia. Evidence-based pharmacological therapies for treating substance use disorders, such as opioid agonist treatments with methadone, are generally unavailable in these settings.
Richard Suu-Ire et al.
PLoS neglected tropical diseases, 12(3), e0006311-e0006311 (2018-03-06)
Rabies is a fatal neurologic disease caused by lyssavirus infection. People are infected through contact with infected animals. The relative increase of human rabies acquired from bats calls for a better understanding of lyssavirus infections in their natural hosts. So

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service