Skip to Content
Merck
All Photos(1)

Key Documents

Y0000506

Cefotaxime for peak identification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Cefotaxime sodium salt, Cefotaxim sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H16N5NaO7S2
CAS Number:
Molecular Weight:
477.45
Beilstein:
5711411
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

cefotaxime

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[Na+].[H][C@]12SCC(COC(C)=O)=C(N1C(=O)[C@H]2NC(=O)C(=N/OC)\c3csc(N)n3)C([O-])=O

InChI

1S/C16H17N5O7S2.Na/c1-6(22)28-3-7-4-29-14-10(13(24)21(14)11(7)15(25)26)19-12(23)9(20-27-2)8-5-30-16(17)18-8;/h5,10,14H,3-4H2,1-2H3,(H2,17,18)(H,19,23)(H,25,26);/q;+1/p-1/b20-9-;/t10-,14-;/m1./s1

InChI key

AZZMGZXNTDTSME-JUZDKLSSSA-M

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Cefotaxime for peak identification EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Cefotaxim inhibits bacterial cell wall synthesis by binding to penicillin-binding proteins (PBPs) which inhibits the final transpeptidation step of peptidoglycan synthesis in bacterial cell walls. As a result, bacteria lyse due to cell wall autolytic enzymes.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Broad spectrum third generation cephalosporin antibiotic.
Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hien Bao Dieu Thai et al.
The Analyst, 140(8), 2804-2809 (2015-02-28)
We described a dual turn-on probe sensitive to both acidity and basicity, which could be designed by connecting a fluorophore to a quencher via metal-ligand interaction. Atto488-labeled nitrilotriacetic acid and polyhistidine peptide were used as the fluorophore and the quencher
R N Jones
Diagnostic microbiology and infectious disease, 13(4), 317-327 (1990-07-01)
Cefotaxime is a parenteral broad-spectrum cephalosporin, used extensively worldwide for chemotherapy of serious infections. Since its release in 1979, cefotaxime has also been studied to minimize surgery-related infections and, more than any other new compound, has been used in a
Siiri Kõljalg et al.
APMIS : acta pathologica, microbiologica, et immunologica Scandinavica, 122(5), 452-458 (2013-09-17)
The aim of our study was to characterize the phylogenetic groups of Escherichia coli, antibiotic resistance, and containment of class 1 integrons in the first attack of pyelonephritis and in subsequent recurrences in young children. Altogether, 89 urine E. coli
Alje P van Dam et al.
Sexually transmitted infections, 90(7), 513-514 (2014-08-13)
We describe the first case of treatment failure of gonorrhoea with a third generation cephalosporin, cefotaxime 1g intramuscularly, in the Netherlands. The case was from a high-frequency transmitting population (men having sex with men) and was caused by the internationally
Fu Peng et al.
Chinese medical journal, 127(23), 4001-4005 (2014-11-29)
Our pervious antibacterial studies on several traditional Chinese medicines have found that Patchouli oil from Pogostemon cablin had significant antibacterial activity against methicillin-resistant Staphylococcus aureus (MRSA), which has spread worldwide and infected innumerable people. In order to find the more

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service