Skip to Content
Merck
All Photos(1)

Key Documents

Y0000466

Povidone (iodinated)

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Poly(vinylpyrrolidone)–Iodine complex, PVP-I, Povadyne® antiseptic

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

povidone

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

II.C=CN1CCCC1=O

InChI

1S/C6H9NO.I2/c1-2-7-5-3-4-6(7)8;1-2/h2H,1,3-5H2;

InChI key

CPKVUHPKYQGHMW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Povidone (iodinated) EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Legal Information

Povadyne is a registered trademark of National Patent Development Corp.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

>212.0 °F - Not applicable

Flash Point(C)

> 100 °C - Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kunal A Lodhia et al.
The Annals of otology, rhinology, and laryngology, 124(1), 68-71 (2014-07-30)
It is common practice to irrigate the operative site following tumor resection during major head and neck surgery. A variety of irrigation solutions are used, but there are few data on their relative efficacies in this context. The effect of
Wound dressing following debridement for Fournier's gangrene.
J M Henderson et al.
Annals of the Royal College of Surgeons of England, 96(4), 311-311 (2014-05-02)
William F Schleicher et al.
Dermatologic surgery : official publication for American Society for Dermatologic Surgery [et al.], 40(10), 1103-1107 (2014-09-18)
Refrigerant sprays have been widely used to reduce pain in the office setting. However, more recently, their use has been limited by both concern regarding flammability and questions of bacterial contamination. We investigated the microbiological effect of 1,1,1,3,3 pentafluoropropane and
F P Schmidt et al.
The Journal of cardiovascular surgery, 55(2), 271-277 (2013-10-18)
Saphenous vein grafts harvested for use as bypass conduits can be contaminated intraoperatively, e.g. by being inadvertently dropped to the floor of the operating room (OR). This study was performed to investigate microorganisms most likely contaminating vein grafts and to
M A Reyazulla et al.
European annals of allergy and clinical immunology, 46(4), 157-159 (2014-07-24)
Povidone iodine (PVP-I) is a chemical complex of polyvinylpyrrolidone (also known as povidone or PVP) and elemental iodine. Iodine containing compounds are widely and commonly used as antiseptics and disinfectants. They are available in various forms like aqueous solution, tincture

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service