Skip to Content
Merck
All Photos(4)

Key Documents

K1637

Sigma-Aldrich

Kanamycin sulfate from Streptomyces kanamyceticus

meets USP testing specifications, powder

Synonym(s):

Kanamycin A, Kanamycin sulfate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H36N4O11 · H2O4S
CAS Number:
Molecular Weight:
582.58
EC Number:
MDL number:
UNSPSC Code:
51281654
PubChem Substance ID:
NACRES:
NA.76

biological source

Streptomyces kanamyceticus

Quality Level

Agency

USP/NF
meets USP testing specifications

form

powder

potency

≥750 μg per mg

solubility

H2O: 10-50 mg/mL (As a stock solution. Stock solutions should be stored at 2-8°C. Stable at 37°C for 5 days.)

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria
mycobacteria
mycoplasma

application(s)

pharmaceutical (small molecule)

Mode of action

protein synthesis | interferes

SMILES string

OS(O)(=O)=O.NC[C@H]1O[C@H](O[C@@H]2[C@@H](N)C[C@@H](N)[C@H](O[C@H]3O[C@H](CO)[C@@H](O)[C@H](N)[C@H]3O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C18H36N4O11.H2O4S/c19-2-6-10(25)12(27)13(28)18(30-6)33-16-5(21)1-4(20)15(14(16)29)32-17-11(26)8(22)9(24)7(3-23)31-17;1-5(2,3)4/h4-18,23-29H,1-3,19-22H2;(H2,1,2,3,4)/t4-,5+,6-,7-,8+,9-,10-,11-,12+,13-,14-,15+,16-,17-,18-;/m1./s1

InChI key

OOYGSFOGFJDDHP-KMCOLRRFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Kanamycin Sulfate is a broad spectrum aminoglycoside-antibiotic purified from the bacterium Streptomyces kanamyceticus. Kanamycin Sulfate is a selection agent for cells in culture media and acts by binding to the 30S subunit of the bacterial ribosome which inhibits protein synthesis in susceptible bacteria.

Application

Kanamycin sulfate is used as an additive in culture media for the isolation of group D streptococci on Kanamycin Esculin Azide Agar and for selection of transformed plant cells containing the neomycin phosphotransferase on a kanamycin-medium. Kanamycin sulfate can also be used as a selection agent for cells transformed with kanamycin B resistance gene. It is recommended for use in cell culture applications at 100 μg/mL. Kanamycin sulfate from Streptomyces kanamyceticus has been used: in the research of cell type-specific expression of genes

Biochem/physiol Actions

Mode of Action: The product acts by binding to the 70S ribosomal subunit, inhibiting translocation and eliciting miscoding.

Mode of Resistance:Aminoglycoside-modifying enzymes (including acetyltransferase, phosphotransferase, nucleotidyltransferase) can alter this antibiotic, preventing its interaction with ribosomes.

Antimicrobial spectrum: Kanamycin sulfate is effective against gram-negative and gram-postiive bacteria, and mycoplasma.

Features and Benefits

  • High quality antibiotic suitable for mulitple research applications
  • meets USP testing specifications

Preparation Note

Kanamycin sulfate is soluble in water at 50 mg/mL, yielding a clear solution. It is practically insoluble in alcohol, acetone, chloroform, ether and ethyl acetate. A 1% solution in water has a pH of 6.5 to 8.5. Sterile solutions can be prepared by a sterile filtration, through a .2μm filter.

Solutions are stable at 37°C for approximately 5 days. Aqueous stock solutions can be stored at 2-8°C for long term storage.

Storage and Stability

Tightly closed. Dry. Keep in a well -ventilated place. Keep locked up or in an area accessible

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Byron H Hartman et al.
Journal of the Association for Research in Otolaryngology : JARO, 10(3), 321-340 (2009-04-18)
The Notch signaling pathway is known to have multiple roles during development of the inner ear. Notch signaling activates transcription of Hes5, a homologue of Drosophila hairy and enhancer of split, which encodes a basic helix-loop-helix transcriptional repressor. Previous studies
Torbjörn Johansson et al.
Genesis (New York, N.Y. : 2000), 48(4), 264-280 (2010-02-10)
Transgenic mice are highly valuable tools for biological research as they allow cell type-specific expression of functionally instrumental genes. In this protocol, the generation of bacterial artificial chromosome (BAC) transgenic constructs is described. We give an overview of different transgenic
Mitsutaka Kitano et al.
Antimicrobial agents and chemotherapy, 58(8), 4795-4803 (2014-06-11)
Highly pathogenic avian influenza A (H5N1) viruses cause severe and often fatal disease in humans. We evaluated the efficacy of repeated intravenous dosing of the neuraminidase inhibitor peramivir against highly pathogenic avian influenza virus A/Vietnam/UT3040/2004 (H5N1) infection in cynomolgus macaques.
Atef Allam et al.
Journal of immunology (Baltimore, Md. : 1950), 193(2), 871-878 (2014-06-11)
The role of the TNF family member CD70 in adaptive T cell responses has been intensively studied, but its function in innate responses is still under investigation. In this study, we show that CD70 inhibits the early innate response to
Fakhri Jeddi et al.
Antimicrobial agents and chemotherapy, 58(8), 4866-4874 (2014-06-11)
Antimonials remain the first-line treatment for the various manifestations of leishmaniasis in most areas where the disease is endemic, and increasing cases of therapeutic failure associated with parasite resistance have been reported. In this study, we assessed the molecular status

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service