Skip to Content
Merck
All Photos(1)

Key Documents

I114

Supelco

p-Iodoclonidine hydrochloride

analytical standard, for drug analysis

Synonym(s):

2-[(2,6-Dichloro-4-iodophenyl)imino]imidazoline hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H8Cl2IN3 · HCl
CAS Number:
Molecular Weight:
392.45
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard, for drug analysis

Quality Level

form

solid

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

color

white to off-white

solubility

H2O: soluble

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

Cl[H].Clc1cc(I)cc(Cl)c1\N=C2/NCCN2

InChI

1S/C9H8Cl2IN3.ClH/c10-6-3-5(12)4-7(11)8(6)15-9-13-1-2-14-9;/h3-4H,1-2H2,(H2,13,14,15);1H

InChI key

ULCGXOSKNHMYAX-UHFFFAOYSA-N

Gene Information

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

High affinity α2 adrenergic receptor agonist.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jan Höcker et al.
The Journal of pharmacy and pharmacology, 61(7), 901-910 (2009-07-11)
alpha(2)-Adrenergic and mu-opioid receptors belong to the rhodopsin family of G-protein coupled receptors and mediate antinociceptive effects via similar signal transduction pathways. Previous studies have revealed direct functional interactions between both receptor systems including synergistic and additive effects. To evaluate
H Zhu et al.
Life sciences, 61(19), 1973-1983 (1997-01-01)
One subtype of imidazoline receptors (IR1) is similar to alpha 2-adrenoceptors (alpha 2 AR) based on their high affinity for clonidine and related imidazoline compounds. On the other hand, IR1 possess low affinity for norepinephrine (NE) and other catecholamines. Imidazoline
N Benedetto et al.
Pathologie-biologie, 45(1), 9-18 (1997-01-01)
We investigated the effects of alpha-adrenergic on the capacity of Toxoplasma gondii to invade and proliferate in cultured human umbilical vein endothelial cells. Pretreatment of human umbilical vein endothelial cells (HUVEC) with alpha 2-adrenergic led to a high degree of
M Van Dort et al.
Journal of medicinal chemistry, 30(7), 1241-1244 (1987-07-01)
The chemical synthesis of 2-[(2,6-dichloro-4-iodophenyl)imino]imidazolidine (PIC) and its radioiodinated analogue [125I]PIC is described. PIC was synthesized from 2,6-dichloroaniline in five synthetic steps. This agent displayed a high affinity for the alpha 2-adrenergic receptor (IC50 = 1.5 nM) in competitive binding
Y Jin et al.
Investigative ophthalmology & visual science, 35(5), 2500-2508 (1994-04-01)
This study sought to identify and characterize subtypes of alpha 2-adrenoceptors in rabbit ciliary body. Radioligand binding assays were performed with the alpha 2-agonist ligand [125I]-p-iodoclonidine ([125I]PIC) and the antagonist ligand [3H]rauwolscine. Both [125I]PIC and [3H]rauwolscine bound to a single

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service