Skip to Content
Merck
All Photos(1)

Key Documents

I0800000

Isotretinoin

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

13-cis-Retinoic acid, Isotretinoin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H28O2
CAS Number:
Molecular Weight:
300.44
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

biological source

synthetic

grade

pharmaceutical primary standard

Agency

EP

API family

isotretinoin

form

solid

manufacturer/tradename

EDQM

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

mp

172-175 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[H]\C(C(O)=O)=C(C)\C=C\C=C(C)\C=C\C1=C(C)CCCC1(C)C

InChI

1S/C20H28O2/c1-15(8-6-9-16(2)14-19(21)22)11-12-18-17(3)10-7-13-20(18,4)5/h6,8-9,11-12,14H,7,10,13H2,1-5H3,(H,21,22)/b9-6+,12-11+,15-8+,16-14-

InChI key

SHGAZHPCJJPHSC-XFYACQKRSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.
Isotretinoin, a vital isomer of vitamin A, is implicated in the treatment of severe recalcitrant nodular acne disorder. It is the only drug known to operate against the major pathogenic factors of acne. It serves to diminish the sebaceous gland activity, thereby resulting in the correction of keratinization defect.

Application

Isotretinoin may be used as an EP reference standard for the determination of the analyte in pharmaceutical formulations by various chromatography techniques.

Biochem/physiol Actions

13-cis-Retinoic acid (RA) has anti-inflammatory and anti-tumor action. The action of RA is mediated through RAR-β and RAR-α receptors. RA attenuates iNOS expression and activity in cytokine-stimulated murine mesangial cells. It induces mitochondrial membrane permeability transition, observed as swelling and as a decrease in membrane potential, and stimulates the release of cytochrome c implicating mechanisms through the apoptosis pathway. These activities are reversed by EGTA and cyclosporin A. RA also increases MMP-1 protein expression partially via increased transcription.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isotretinoin
European Pharmacopoeia Commission and European Directorate for the Quality of Medicines & Healthcare
European pharmacopoeia, 3(5), 5705-5706 (2018)
Determination of isotretinoin in pharmaceutical formulations by reversed-phase HPLC
Guimar?es CA, et al.
Journal of Biomedical Science and Engineering, 3(5), 454-458 (2010)
Development of a gas chromatography method for the determination of isotretinoin and its degradation products in pharmaceuticals
Lima EM, et al.
Journal of Pharmaceutical and Biomedical Analysis, 38(4), 678-685 (2005)
Emily Yiping Gan et al.
The Journal of dermatological treatment, 24(5), 387-391 (2012-03-07)
Oral isotretinoin is effective for acne vulgaris but concerns remain regarding its adverse effects. This study aims to evaluate the safety and efficacy of isotretinoin for acne vulgaris in Asian patients. We conducted a 4-year retrospective study on all patients
Mahyar Etminan et al.
JAMA dermatology, 149(2), 216-220 (2013-02-22)
To examine the association between isotretinoin and the risk for inflammatory bowel disease (IBD) among women of reproductive age. Nested case-control study and meta-analysis. A US health claims database. We formed a cohort of women aged 18 to 46 years

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service