Skip to Content
Merck
All Photos(1)

Key Documents

32932

Supelco

Neosolaniol

Synonym(s):

Neosolaniol from Fusarium sp., 4β,15-Diacetoxy-3α,8α-dihydroxy-12,13-epoxytrichothec-9-ene, 8-Hydroxydiacetoxyscirpenol, NEO

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H26O8
CAS Number:
Molecular Weight:
382.40
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard
reference material

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

−20°C

SMILES string

CC(=O)OC[C@]12C[C@H](O)C(C)=C[C@H]1O[C@@H]3[C@H](O)[C@@H](OC(C)=O)[C@@]2(C)[C@]34CO4

InChI

1S/C19H26O8/c1-9-5-13-18(6-12(9)22,7-24-10(2)20)17(4)15(26-11(3)21)14(23)16(27-13)19(17)8-25-19/h5,12-16,22-23H,6-8H2,1-4H3/t12-,13+,14+,15+,16+,17+,18+,19-/m0/s1

InChI key

TVZHDVCTOCZDNE-WVJYZQHISA-N

Looking for similar products? Visit Product Comparison Guide

General description

Neosolaniol belongs to the class of type A trichothecene mycotoxins produced by various Fusarium molds.

Application

Neosolaniol may be used as an analytical reference standard for the determination of the analyte in:
  • Wheat samples by acetonitrile-based extraction and gas chromatography-tandem mass spectrometry (GC-MS/MS) as well as capillary GC with flame ionization detection (FID).
  • Cereals by acetonitrile-based extraction and GC coupled to mass spectrometry (MS).

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Dongmei Jiang et al.
Toxins, 10(10) (2018-10-13)
One-step solid-phase extraction (SPE) using a multiwalled carbon nanotube (MWCNT) for simultaneous analysis of 21 mycotoxins, including nine trichothecenes, zearalenone (ZEN) and its derivatives, four aflatoxins, and two ochratoxins, in corn and wheat was developed. Several key parameters affecting the
Determination of trichothecenes in wheat by capillary gas chromatography with flame ionisation detection.
Schothorst RC and Jekel AA
Food Chemistry, 73(1), 111-117 (2001)
Yijia Deng et al.
Drug and chemical toxicology, 41(1), 113-122 (2017-05-10)
T-2 toxin (T-2) is a type-A trichothecene produced by Fusarium that causes toxicity to animals. T-2 contamination of grain-based aquatic feed is a concern for the industries related to edible aquatic crustacean species such as the shrimp industry because it
Lorenzo De Colli et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1150, 122187-122187 (2020-05-31)
A method was developed and validated for the simultaneous determination of 42 mycotoxins in oats. The method includes all the mycotoxins listed under Commission Regulation 1881/2006 and Commission Recommendation 165/2013, the emerging mycotoxins (beauvericin, alternariol, alternariol-methyl-ether and enniatins), and two
Kraiwut Nualkaw et al.
Toxins, 12(4) (2020-04-17)
A reliable, sensitive and accurate multiple mycotoxin method was developed for the simultaneous determination of 17 mycotoxins in swine, poultry and dairy feeds using stable isotope dilution (13C-ISTD) and (ultra)-high performance liquid chromatography-tandem mass spectrometry (HPLC-MS/MS). A simple QuEChERS-based method

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service