Skip to Content
Merck
All Photos(1)

Key Documents

11373242910

Roche

Fluorescein-12-dUTP

≥85% (HPLC), suitable for ELISA, suitable for hybridization, solution

Synonym(s):

Fluorescein-12-dUTP, dUTP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116100

Quality Level

Assay

≥85% (HPLC)

form

solution

mol wt

Mr 1018.4 (fluorescein-12-dUTP-Li4)

packaging

pkg of 25 μL (25 nmol; 1mM)

manufacturer/tradename

Roche

technique(s)

ELISA: suitable
hybridization: suitable

storage temp.

−20°C

Application

Fluorescein-12-dUTP is used as a substrate for:
  • terminal transferase
  • DNA polymerase I (holoenzyme and Klenow fragment),
  • Taq DNA polymerase
  • reverse transcriptase (e.g., from AMV and M-MuLV).
Fluorescein-12-dUTP has been used to label nuclear extracts during PCR. It has also been used to replace dTTP in the random-primed DNA labeling reaction or in nick translation reactions, as well as in PCR. The nucleotide also serves as a substrate for terminal transferase in 3′-end labeling and TUNEL. Fluorescein-labeled probes can be used for in in situ hybridization with direct fluorescence detection or detection by enzyme linked immunoimmunsorbent assay (ELISA) using Anti-Fluorescein-AP, Fab fragments. Repeated fluorescence labeling using Tetramethyl-Rhodamine-6-dUTP (red) and AMCA-6-dUTP (bright blue) is possible.

Quality

Typical analysis: At least 85% Fluorescein-12-dUTP (HPLC, area%).

Physical form

Fluorescein-12-dUTP, tetralithium salt.

Storage and Stability

The unopened reagent is stable at 15 to 25°C until the expiration date printed on the label. Decomposition of approx. 5% may occur within 6 months.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Pro-survival redox signalling in progesterone-mediated retinal
neuroprotection
Lopez R, et al.
The European Journal of Neuroscience, 46(1) (2017)
Nucleoporin 107, 62 and 153 mediate Kcnq1ot1
imprinted domain regulation in extraembryonic
endoderm stem cells
Sachani SS, et al.
Nature Communications (2018)
Shang-mian Yie et al.
Human reproduction (Oxford, England), 21(10), 2538-2544 (2006-05-11)
We have previously demonstrated that progesterone has a stimulatory effect on HLA-G gene expression. Because this effect was abolished by the anti-progestin, RU486, we hypothesize that this effect is through receptor-mediated up-regulation of the HLA-G gene. The objective of this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service