Skip to Content
Merck
All Photos(1)

Key Documents

1.07253

Supelco

Phenylhydrazinium chloride

for analysis

Synonym(s):

Phenylhydrazinium chloride, Phenylhydrazine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H9ClN2
CAS Number:
Molecular Weight:
144.60
MDL number:
UNSPSC Code:
12352107
EC Index Number:
200-444-7
NACRES:
NB.21

Agency

reag. Ph. Eur.

Quality Level

Assay

≥99.0% (silver nitrate titration)

form

solid

sulfated ash

≤0.1%

loss

≤0.5% loss on drying, 105 °C

pH

2.6-2.9 (50 g/L in H2O)

mp

245 °C (decomposition)

solubility

50 g/L

bulk density

330 kg/m3

anion traces

sulfate (SO42-): ≤0.01%

cation traces

Fe: ≤0.001%

storage temp.

2-30°C

InChI

1S/C6H8N2.ClH/c7-8-6-4-2-1-3-5-6;/h1-5,8H,7H2;1H

InChI key

JOVOSQBPPZZESK-UHFFFAOYSA-N

Analysis Note

Assay (argentometric): ≥ 99.0 %
Identity (IR): passes test
Solubility in ethanol: passes test
Solubility in water: passes test
Sulfate (SO₄): ≤ 0.01 %
Iron (Fe): ≤ 0.001 %
Sulfated ash: ≤ 0.1 %
Loss on drying (105 °C): ≤ 0.5 %

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Carc. 1B - Eye Irrit. 2 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service