Skip to Content
Merck
All Photos(1)

Key Documents

C-094

Supelco

Carbamazepine-D10 solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15D10H2N2O
CAS Number:
Molecular Weight:
246.33
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

[2H]C1=C2N(C(N)=O)C3=C(C([2H])=C([2H])C([2H])=C3[2H])C([2H])=C([2H])C2=C([2H])C([2H])=C1[2H]

InChI

1S/C15H12N2O/c16-15(18)17-13-7-3-1-5-11(13)9-10-12-6-2-4-8-14(12)17/h1-10H,(H2,16,18)/i1D,2D,3D,4D,5D,6D,7D,8D,9D,10D

InChI key

FFGPTBGBLSHEPO-LHNTUAQVSA-N

General description

Carbamazepine is an anticonvulsant and mood stabilizing drug used primarily in the treatment of epilepsy and neuropathic pain. This stable-labeled internal standard is suitable for use in quantitation of carbamazepine levels or isotope dilution methods by GC/MS or LC/MS for applications in pain prescription monitoring, clinical toxicology, therapeutic drug monitoring or urine drug testing.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

The application note describes a comparative analysis of plasma protein binding results for free/bound drugs using two different sample extraction steps, with and without agitation.

The application note describes a comparative analysis of plasma protein binding results for free/bound drugs using two different sample extraction steps, with and without agitation.

The application note describes a comparative analysis of plasma protein binding results for free/bound drugs using two different sample extraction steps, with and without agitation.

The application note describes a comparative analysis of plasma protein binding results for free/bound drugs using two different sample extraction steps, with and without agitation.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service