Skip to Content
Merck
All Photos(2)

Key Documents

850376C

Avanti

18:1 (Δ9-Trans) PC

1,2-dielaidoyl-sn-glycero-3-phosphocholine, chloroform

Synonym(s):

1,2-di-(9E-octadecenoyl)-sn-glycero-3-phosphocholine; PC(18:1(9E)/18:1(9E))

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H84NO8P
CAS Number:
Molecular Weight:
786.11
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (850376C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850376C

concentration

10 mg/mL (850376C-25mg)

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

InChI

1S/C44H84NO8P/c1-6-8-10-12-14-16-18-20-22-24-26-28-30-32-34-36-43(46)50-40-42(41-52-54(48,49)51-39-38-45(3,4)5)53-44(47)37-35-33-31-29-27-25-23-21-19-17-15-13-11-9-7-2/h20-23,42H,6-19,24-41H2,1-5H3/b22-20+,23-21+

InChI key

SNKAWJBJQDLSFF-DQPVQCHKSA-N

General description

18:1 (Δ9-Trans) PC is a phosphoglycerolipid, which consists of two unsaturated acyl chains.
The list of Phosphatidylcholine products offered by Avanti is designed to provide compounds having a variety of physical properties. Products available include short chain (C3-C8 are water soluble and hygroscopic), saturated, multi-unsaturated and mixed acid PC′s. All of the products are purified by HPLC, and special precautions are taken to protect the products for oxidization and hydrolysis.

Application

18:1 (Δ9-Trans) PC is suitable for use as a synthetic lipid to study its effects on sarcoplasmic reticulum (SR) Ca2+-ATPase (SERCA) activity.

Packaging

5 mL Clear Glass Sealed Ampule (850376C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Charge transfer dissociation of phosphocholines: gas-phase ion/ion reactions between helium cations and phospholipid cations
Li P and Jackson GP
Journal of Mass Spectrometry : Jms, 52(5), 271-282 (2017)
Radical-induced fragmentation of phospholipid cations using metastable atom-activated dissociation mass spectrometry (MAD-MS)
Deimler RE, et al.
International Journal of Mass Spectrometry, 390, 178-186 (2015)
Activating and deactivating roles of lipid bilayers on the Ca2+-ATPase/phospholamban complex
Gustavsson M, et al.
Biochemistry, 50(47), 10367-10374 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service