Skip to Content
Merck
All Photos(1)

Key Documents

Z742477

Optional tube holder for BeadBlaster

Benchmark Scientific D2400-R5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.61

feature

Holds 12 x 5 mL tubes

packaging

pack of 1 ea

manufacturer/tradename

Benchmark Scientific D2400-R5

technique(s)

PCR: suitable

General description

5ml Tube Holder for D2400

Legal Information

BeadBlaster is a trademark of Benchmark Scientific, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service