Key Documents
Z677418
Dermatril P nitrile gloves
size XL
About This Item
UNSPSC Code:
46181504
NACRES:
NB.35
material
blue gloves
blue nitrile
size
XL
sterility
non-sterile
feature
CE compliant
disposable
latex free
powder-free
packaging
pack of 50 ea (boxes of 50)
manufacturer/tradename
KCL 743 - 10
KCL 743
L × thickness
270 mm × 0.2 mm
General description
Dermatril P is an extended cuff, blue nitrile, powder-free glove providing a high level of protection against chemical and bacteriological contamination. The Dermatril P disposable glove also displays very good physical durability in comparison to conventional disposable gloves.
All Dermatril P disposable gloves are INDIVIDUALLY marked with the appropriate standards mark applied on the inside using a food-friendly dye.
Features and Benefits
  • Anti-static
  • Free from allergenic substances such as thiurames, benzothiazoles and thiourea
  • Roughened fingertips for better grip
  • Rolled cuff edge for easier donning
  • CE compliant
  • Meet standards: EN 374 (cat. III), EN 388 (cat. II), EN 61340-5-1, ASTM 1671:2001
Legal Information
Dermatril is a trademark of KCL GmbH