Skip to Content
Merck
All Photos(2)

Key Documents

699152

Sigma-Aldrich

Bathocuproine

greener alternative

sublimed grade, 99.99% trace metals basis

Synonym(s):

2,9-Dimethyl-4,7-diphenyl-1,10-phenanthroline, BCP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H20N2
CAS Number:
Molecular Weight:
360.45
Beilstein:
306714
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

grade

sublimed grade

Quality Level

Assay

99.99% trace metals basis

form

powder

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

279-283 °C (lit.)
285 °C (DSC)

λmax

277 nm

fluorescence

λem 386 nm in THF

Orbital energy

HOMO 7 eV 
LUMO 3.5 eV 

OLED Device Performance

ITO/CuPc/NPD/TCTA/mCP:Firpic (6%)/BCP/LiF/Al

  • Color: blue
  • Max. EQE: 4.2 %

ITO/NPD/CBP:Ir(ppy)3/BCP/Alq3/Mg:Al
  • Color: green
  • Max. Luminance: 100000 Cd/m2
  • Max. EQE: 8 %
  • Turn-On Voltage: 4.3 V

ITO/NPD/TCTA/BCPO:Ir(piq)3 (7-8%)/BCP/Alq3/LiF/Al
  • Color: red
  • Max. Luminance: 24529 Cd/m2
  • Max. EQE: 17 %
  • Turn-On Voltage: 2.7 V

ITO/NPD/TCTA/BCPO:Ir(ppy)3 (7-8%)/BCP/Alq3/LiF/Al
  • Color: green
  • Max. Luminance: 207839 Cd/m2
  • Max. EQE: 21.6 %
  • Turn-On Voltage: 2.1 V

greener alternative category

SMILES string

Cc1cc(-c2ccccc2)c3ccc4c(cc(C)nc4c3n1)-c5ccccc5

InChI

1S/C26H20N2/c1-17-15-23(19-9-5-3-6-10-19)21-13-14-22-24(20-11-7-4-8-12-20)16-18(2)28-26(22)25(21)27-17/h3-16H,1-2H3

InChI key

STTGYIUESPWXOW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

TGA/DSC Lot specific scans available upon request
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product belongs to Enabling category of greener alternatives thus aligns with "Design for energy efficency". Electron transport organic materials have the chemical functionality to transport electrons and are used as electron transport layer in OLED devices. Click here for more information.

Application

Organic electronic material useful as OLED electron transporter and hole blocker.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joab E C Hwang et al.
Metallomics : integrated biometal science, 6(12), 2223-2229 (2014-10-18)
The human copper homeostasis disorders Menkes and Wilson disease both have severe neurological symptoms. Menkes is a copper deficiency disorder whereas Wilson disease patients suffer from copper toxicity, indicating that tight control of neuronal copper levels is essential for proper
Malin Hultberg et al.
Chemico-biological interactions, 163(3), 192-198 (2006-09-15)
Glutathione plays crucial roles in antioxidant defence and glutathione deficiency contributes to oxidative stress and may therefore play a key role in the pathogenesis of many diseases. The objectives of the present study were to evaluate the effects on glutathione
Wendy A Spencer et al.
Free radical biology & medicine, 46(10), 1346-1352 (2009-02-24)
Polychlorinated biphenyls (PCBs) are toxic industrial chemicals, complete carcinogens, and efficacious tumor promoters. However, the mechanism(s) of PCB-mediated carcinogenicity remains largely undefined. One likely pathway by which these agents may play a role in carcinogenesis is the generation of oxidative
B M Krenn et al.
Journal of virology, 79(22), 13892-13899 (2005-10-29)
Pyrrolidine dithiocarbamate (PDTC) is an antiviral compound that was shown to inhibit the replication of human rhinoviruses (HRVs), poliovirus, and influenza virus. To elucidate the mechanism of PDTC, the effects on the individual steps of the infection cycle of HRV
Kaoru Midorikawa et al.
Chemico-biological interactions, 150(3), 271-281 (2004-11-25)
Ethylbenzene is carcinogenic to rats and mice, while it has no mutagenic activity. We have investigated whether ethylbenzene undergoes metabolic activation, leading to DNA damage. Ethylbenzene was metabolized to 1-phenylethanol, acetophenone, 2-ethylphenol and 4-ethylphenol by rat liver microsomes. Furthermore, 2-ethylphenol

Articles

Silylethyne substitution offers an opportunity to tune solubility for application-specific needs and self-assembly for electronic performance and has yielded semiconductors with excellent device performance.

Silylethyne substitution offers an opportunity to tune solubility for application-specific needs and self-assembly for electronic performance and has yielded semiconductors with excellent device performance.

Silylethyne substitution offers an opportunity to tune solubility for application-specific needs and self-assembly for electronic performance and has yielded semiconductors with excellent device performance.

Silylethyne substitution offers an opportunity to tune solubility for application-specific needs and self-assembly for electronic performance and has yielded semiconductors with excellent device performance.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service