Skip to Content
Merck
All Photos(1)

Key Documents

565415

Sigma-Aldrich

Diethyl allylphosphonate

98%

Synonym(s):

2-Propenyl-phosphonic acid diethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCH2P(O)(OC2H5)2
CAS Number:
Molecular Weight:
178.17
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

reaction suitability

reaction type: C-C Bond Formation

refractive index

n20/D 1.4340 (lit.)

bp

46 °C/0.35 mmHg (lit.)

bulk density

1.022 g/mL

SMILES string

CCOP(=O)(CC=C)OCC

InChI

1S/C7H15O3P/c1-4-7-11(8,9-5-2)10-6-3/h4H,1,5-7H2,2-3H3

InChI key

YPJHXRAHMUKXAE-UHFFFAOYSA-N

Application

Reactant for:
  • Copolymerization of phosphonated allyl monomers and maleic anhydride
  • Enantioselective synthesis of solamin type mono-THF acetogenins
  • RCM reaction yielding oxaphospholene and oxaphosphinene heterocycles
  • Synthesis of spongistatin 2 using Wittig coupling
  • Stereoselective synthesis of pentacyclic furanosteroids
  • Preparation of protected polyhydroxylated β -amino acid constitutents of microsclerodermins

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Elizabeth I Parkinson et al.
Nature chemical biology, 15(11), 1049-1056 (2019-08-28)
Fosmidomycin and related molecules comprise a family of phosphonate natural products with potent antibacterial, antimalarial and herbicidal activities. To understand the biosynthesis of these compounds, we characterized the fosmidomycin producer, Streptomyces lavendulae, using biochemical and genetic approaches. We were unable

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service