Skip to Content
Merck
All Photos(2)

Key Documents

389153

Sigma-Aldrich

β-Cyclodextrin, sulfated sodium salt

extent of labeling: 12-15 mol per mol β-CD

Synonym(s):

β-Cyclodextrin, hydrogen sulfate sodium salt, Sodium salt of sulfated cycloheptaamylose

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.22

form

powder

autoignition temp.

770 °F

extent of labeling

12-15 mol per mol β-CD

InChI

1S/C42H70O56S7/c43-15-22(50)36-85-8(1-78-99(57,58)59)29(15)92-37-23(51)16(44)31(10(86-37)3-80-101(63,64)65)94-39-25(53)18(46)33(12(88-39)5-82-103(69,70)71)96-41-27(55)20(48)35(14(90-41)7-84-105(75,76)77)98-42-28(56)21(49)34(13(91-42)6-83-104(72,73)74)97-40-26(54)19(47)32(11(89-40)4-81-102(66,67)68)95-38-24(52)17(45)30(93-36)9(87-38)2-79-100(60,61)62/h8-56H,1-7H2,(H,57,58,59)(H,60,61,62)(H,63,64,65)(H,66,67,68)(H,69,70,71)(H,72,73,74)(H,75,76,77)/t8-,9-,10-,11-,12-,13-,14-,15-,16-,17-,18-,19-,20-,21-,22-,23-,24-,25-,26-,27-,28-,29-,30-,31-,32-,33-,34-,35-,36-,37-,38-,39-,40-,41-,42-/m1/s1

InChI key

JFDDAIKYFLAHSD-FOUAGVGXSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Analytic reagent for enantioselective capillary electrophoresis assays for propafenone and its metabolites

Investigations into the effect on fluorescence of aminonaphthalimide dyes

Reactant or reagent for:
  • Complexation of Ru(II) trisdiimine complexes
  • Stereoselective sodium borohydride reduction of cyclohexanone derivatives

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Petra Švecová et al.
Talanta, 198, 154-158 (2019-03-17)
The aim of this study was to focus on the reduction of chiral selector concentration, sulfated-β-cyclodextrin, in an attempt decrease the running costs associated with separating cetirizine enantiomers by capillary electrophoresis. The decrease in the concentration of chiral selector was
Jitka Caslavska et al.
Electrophoresis, 41(7-8), 502-513 (2019-11-09)
For separation of enantiomers in presence of a chiral selector, data obtained with the 1D dynamic simulators SIMUL5complex and GENTRANS are compared to data predicted by PeakMaster 6, a recently released generalized model of the linear theory of electromigration. Four

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service