Skip to Content
Merck
All Photos(1)

Key Documents

269433

Sigma-Aldrich

2,2,3,3,4,4,5,5-Octafluoro-1-pentanol

98%

Synonym(s):

1,1,2,2,3,3,4,4-Octafluoropentan-5-ol, 1,1,5-Trihydro-1-hydroxyperfluoropentane, 1,1,5-Trihydrooctafluoropentan-1-ol, 1,1,5-Trihydrooctafluoropentyl alcohol, 1,1,5-Trihydroperfluoropentan-1-ol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F2CH(CF2)3CH2OH
CAS Number:
Molecular Weight:
232.07
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.318 (lit.)

bp

141-142 °C (lit.)

density

1.667 g/mL at 25 °C (lit.)

SMILES string

OCC(F)(F)C(F)(F)C(F)(F)C(F)F

InChI

1S/C5H4F8O/c6-2(7)4(10,11)5(12,13)3(8,9)1-14/h2,14H,1H2

InChI key

JUGSKHLZINSXPQ-UHFFFAOYSA-N

Application

2,2,3,3,4,4,5,5-Octafluoro-1-pentanol was used as a cosurfactant in the synthesis of silver and silver iodide nanocrystals. It was also used in the synthesis of Ag2S nanocrystals with a characteristic surface plasmon resonance absorption at 330nm.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

168.8 °F - closed cup

Flash Point(C)

76 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juncheng Liu et al.
Chemical communications (Cambridge, England), (22)(22), 2582-2583 (2004-11-16)
Ag2S nanocrystals with a mean diameter of 5.9 nm (sigma= 1.65 nm) and characteristic surface plasmon resonance absorption at 330 nm have been synthesized in water-in-supercritical CO2 reverse microemulsion using the commonly used AOT surfactant with 2,2,3,3,4,4,5,5-octafluoro-1-pentanol (F-pentanol) as cosurfactant.
Juncheng Liu et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 11(6), 1854-1860 (2005-02-03)
Silver and silver iodide nanocrystals have been synthesized in the water-in-CO(2) reverse microemulsions formed by the commonly used surfactant, sodium bis(2-ethylhexyl)sulfosuccinate (AOT), in the presence of 2,2,3,3,4,4,5,5-octafluoro-1-pentanol as cosurfactant. The nanometer-sized aqueous domains in the microemulsion cores not only act
Li-Chong Xu et al.
Journal of biomedical materials research. Part B, Applied biomaterials, 108(8), 3250-3260 (2020-06-20)
Biomaterial-associated microbial infection and thrombosis represent major issues to the success of long-term use of implantable blood-contacting medical devices. The development of new poly[bis(octafluoropentoxy) phosphazene (OFP) biomaterials provides new routes for combatting microbial infection and thrombosis. However, the limited mechanical
Meixian Tang et al.
Bioactive materials, 6(2), 447-459 (2020-10-01)
The utilization of biomaterials in implanted blood-contacting medical devices often induces a persistent problem of microbial infection, which results from bacterial adhesion and biofilm formation on the surface of biomaterials. In this research, we developed new fluorinated alkoxyphosphazene materials, specifically

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service