Skip to Content
Merck
All Photos(2)

Key Documents

235393

Sigma-Aldrich

2,4,6-Trichloroanisole

99%

Synonym(s):

2,4,6-Trichloro-1-methoxybenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl3C6H2OCH3
CAS Number:
Molecular Weight:
211.47
Beilstein:
2329219
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

bp

132 °C/28 mmHg (lit.)

mp

60-62 °C (lit.)

solubility

benzene: soluble(lit.)
cyclohexanone: soluble(lit.)
dioxane: soluble(lit.)
methanol: soluble(lit.)
water: insoluble (practically)(lit.)

SMILES string

COc1c(Cl)cc(Cl)cc1Cl

InChI

1S/C7H5Cl3O/c1-11-7-5(9)2-4(8)3-6(7)10/h2-3H,1H3

InChI key

WCVOGSZTONGSQY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,4,6-Trichloroanisole (TCA) is responsible for the cork taint in wines. TCA has been quantitated in bark cork stoppers by supercritical fluid extraction (SFE) method. Degradation of TCA catalyzed by raw bauxite via ozonation in water has been investigated.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fei Qi et al.
Journal of hazardous materials, 168(1), 246-252 (2009-03-11)
A kind of inexpensive and environmental friendly mineral, the raw bauxite has been used successfully as a catalyst combined with ozonation in the degradation of 2,4,6-trichloroanisole (TCA). The catalyst was characterized by using various analytical techniques. X-ray powder diffraction (XRD)
Elisabetta Pittari et al.
Foods (Basel, Switzerland), 9(11) (2020-10-30)
This work aimed at investigating red wine olfactory-oral cross-modal interactions, and at testing their impact on the correlations between sensory and chemical variables. Seventy-four Italian red whole wines (WWs) from 10 varieties, and corresponding deodorized wines (DWs), were evaluated by
M K Taylor et al.
Journal of agricultural and food chemistry, 48(6), 2208-2211 (2000-07-11)
2,4,6-Trichloroanisole (TCA) is the compound most often associated with cork taint in wines and has been shown to have a very low sensory threshold ( approximately 5 ng/L in wine). A supercritical fluid extraction (SFE) method for TCA in bark

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service