Skip to Content
Merck
All Photos(2)

Key Documents

160393

Sigma-Aldrich

Primaquine bisphosphate

98%

Synonym(s):

8-(4-Amino-1-methylbutylamino)-6-methoxyquinoline diphosphate salt, Primaquine diphosphate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H21N3O · 2H3PO4
CAS Number:
Molecular Weight:
455.34
Beilstein:
3812133
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

mp

205-206 °C (dec.) (lit.)

solubility

water: soluble 50 mg/mL, clear, orange to red

SMILES string

OP(O)(O)=O.OP(O)(O)=O.COc1cc(NC(C)CCCN)c2ncccc2c1

InChI

1S/C15H21N3O.2H3O4P/c1-11(5-3-7-16)18-14-10-13(19-2)9-12-6-4-8-17-15(12)14;2*1-5(2,3)4/h4,6,8-11,18H,3,5,7,16H2,1-2H3;2*(H3,1,2,3,4)

InChI key

GJOHLWZHWQUKAU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Primaquine bisphosphate was used in the colorimetric microplate-based, room temperature assay to determine the urea synthesis in cell culture. It was also used in the preparation of primaquine, an established antiplasmodial drug.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Muta. 2 - Repr. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N P Dhammika Nanayakkara et al.
Antimicrobial agents and chemotherapy, 52(6), 2130-2137 (2008-04-02)
8-Aminoquinolines are an important class of antiparasitic agents, with broad utility and excellent efficacy, but also limitations due to hematological toxicities, primarily methemoglobinemia and hemolysis. One representative from this class, (+/-)-8-[(4-amino-1-methylbutyl)amino]-6-methoxy-4-methyl-5-[3,4-dichlorophenoxy]quinoline succinate (NPC1161C), proved extremely efficacious in animal models of
Robert J X Zawada et al.
Biochemistry and cell biology = Biochimie et biologie cellulaire, 87(3), 541-544 (2009-05-19)
Urea is the major nitrogenous end product of protein metabolism in mammals. Here, we describe a quantitative, sensitive method for urea determination using a modified Jung reagent. This assay is specific for urea and is unaffected by ammonia, a common
Rosalind E Howes et al.
Advances in parasitology, 81, 133-201 (2013-02-07)
Glucose-6-phosphate dehydrogenase (G6PD) is a potentially pathogenic inherited enzyme abnormality and, similar to other human red blood cell polymorphisms, is particularly prevalent in historically malaria endemic countries. The spatial extent of Plasmodium vivax malaria overlaps widely with that of G6PD
Nicholas J White
The Lancet. Infectious diseases, 13(2), 175-181 (2012-11-28)
Falciparum malaria is transmitted by anopheline mosquitoes that have fed on blood containing gametocytes of Plasmodium falciparum. In areas of low malaria transmission, where symptomatic infections contribute substantially to malaria transmission, the use of gametocytocidal drugs reduces the incidence of
Melanie Lödige et al.
International journal of medical microbiology : IJMM, 303(8), 539-547 (2013-09-03)
We present a new class of hybrid molecules consisting of the established antiplasmodial drugs primaquine and chloroquine. No drug is known to date that acts comparably against all stages of Plasmodium in its life cycle. Starting from available precursors, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service