Skip to Content
Merck
All Photos(1)

Key Documents

129852

Sigma-Aldrich

Methyl 2-furoate

98%

Synonym(s):

Methyl furan-2-carboxylate, Methyl pyromucate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6O3
CAS Number:
Molecular Weight:
126.11
Beilstein:
111110
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.487 (lit.)

bp

181 °C (lit.)

density

1.179 g/mL at 25 °C (lit.)

SMILES string

COC(=O)c1ccco1

InChI

1S/C6H6O3/c1-8-6(7)5-3-2-4-9-5/h2-4H,1H3

InChI key

HDJLSECJEQSPKW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Methyl 2-furoate (Methyl furan-2-carboxylate, Methyl pyromucate) was used to synthesize cis-fused 5-oxofuro[2,3-b]furans. It was also used in the preparation of sesquiterpene lactone.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

163.4 °F - closed cup

Flash Point(C)

73 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andreas Kreuzer et al.
Organic letters, 15(13), 3420-3423 (2013-06-26)
The first total synthesis of either enantiomer of Arteludovicinolide A and their biological evaluation is reported, featuring a new strategy for the asymmetric construction of γ-butyrolactones with stereogenic side chains in the 4-position. Starting from the renewable resource methyl 2-furoate
Roland Weisser et al.
Organic letters, 7(24), 5353-5356 (2005-11-18)
[structure: see text] A short and enantioselective synthesis of cis-fused 5-oxofuro[2,3-b]furans, being found in many spongiane diterpenoid natural products, is reported starting from inexpensive methyl 2-furoate. Moreover, the acid-catalyzed rearrangement of the furo[2,3-b]furan framework A to B is observed for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service