Skip to Content
Merck
All Photos(2)

Key Documents

109819

Sigma-Aldrich

Propylamine

98%

Synonym(s):

1-Aminopropane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2NH2
CAS Number:
Molecular Weight:
59.11
Beilstein:
1098243
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39030103
PubChem Substance ID:
NACRES:
NA.22

vapor density

2 (vs air)

vapor pressure

4.79 psi ( 20 °C)

Assay

98%

autoignition temp.

604 °F

expl. lim.

10.4 %

refractive index

n20/D 1.388 (lit.)

bp

48 °C (lit.)

mp

−83 °C (lit.)

solubility

H2O: miscible
alcohol: miscible
diethyl ether: miscible

density

0.719 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCCN

InChI

1S/C3H9N/c1-2-3-4/h2-4H2,1H3

InChI key

WGYKZJWCGVVSQN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Propylamine has been used in the synthesis of single crystals of siliceous ferrierite.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

<-31.0 °F

Flash Point(C)

< -35 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and characterisation of a linear [Cu (pyr) 2] 2+ complex in siliceous ferrierite.
Journal of Materials Chemistry, 10(9), 2109-2113 (2000)
Hernán Ariel Castro et al.
Environmental science and pollution research international, 25(22), 21403-21410 (2017-03-25)
Polystyrene divinylbenzene-based ion exchange resins are employed extensively within nuclear power plants (NPPs) and research reactors for purification and chemical control of the cooling water system. To maintain the highest possible water quality, the resins are regularly replaced as they
Mackenzie G Williams et al.
Langmuir : the ACS journal of surfaces and colloids, 33(5), 1121-1131 (2017-02-09)
The covalent attachment of nonfunctionalized and carboxylic acid-functionalized carbon nanotubes to amine-terminated organic monolayers on gold and silicon surfaces is investigated. It is well established that the condensation reaction between a carboxylic acid and an amine is a viable method
Yingjie Zhou et al.
Dalton transactions (Cambridge, England : 2003), 46(38), 13126-13134 (2017-09-26)
Ionic liquids (ILs) have attracted significant attention because of their unique chemical and physical properties. Immobilization of ILs on solid materials helps increase their efficiency and facilitate their recycling. In our present work, ILs were immobilized on α-zirconium phosphate (ZrP)
Chaoqun Wu et al.
Nanomaterials (Basel, Switzerland), 9(5) (2019-05-10)
Zwitterionic polymers are suitable for replacing poly(ethylene glycol) (PEG) polymers because of their better antifouling properties, but zwitterionic polymers have poor mechanical properties, strong water absorption, and their homopolymers should not be used directly. To solve these problems, a reversible-addition

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service