Sample Preparation & Gel Electrophoresis Troubleshooting
Electric currents, wires, leads, combs, leaks… so many opportunities for trouble. But we still use gels, because electrophoresis remains an effective way to separate proteins — so that the results of antibody-based immunodetection can be fairly unequivocal.
Click on the Sample Preparation & Gel Electrophoresis topics to read about the possible causes and remedies:
- No Bands or Gel Front
- Sample Doesn't Sink to the Bottom of the Well
- Sample Leaking Out of Well
- Bands Are Smeared Vertically
- Too Many Bands
- Gel Running Unusually Slowly
- Gel Running Unusually Fast
- Protein Bands Too Close Together (Not Completely Resolved)
- Leftmost and/or Rightmost Bands of the Gel are Distorted
- Bands Form Smiling Shapes
No Bands or Gel Front | |
---|---|
Possible Cause | Remedy |
Proteins have run off the gel |
|
Not enough protein was loaded on the gel |
|
Sample has diffused away from the well prior to applying power |
|
Not enough SDS in the sample |
|
Sample Doesn't Sink to the Bottom of the Well | |
---|---|
Possible Cause | Remedy |
Not enough glycerol in sample |
|
Sample Leaking Out of Well | |
---|---|
Possible Cause | Remedy |
Wells damaged during comb removal |
|
Wells damaged during sample loading |
|
Bands are Smeared Vertically | |
---|---|
Possible Cause | Remedy |
Incomplete protein solubility blocks proteins from entering the gel |
|
Protein degradation |
|
Too much protein loaded ![]() Effect of loading too much protein on SDS-PAGE (rightmost lane) |
|
Gel run too fast |
|
Poor quality or old gel |
|
Too Many Bands | |
---|---|
Possible Cause | Remedy |
Protein degradation |
|
Protein aggregation |
|
Gel Running Unusually Slowly | |
---|---|
Possible Cause | Remedy |
Buffers too concentrated |
|
Current too low |
|
Gel Running Unusually Fast | |
---|---|
Possible Cause | Remedy |
Buffers too dilute |
|
Current too high |
|
Protein Bands Too Close Together (Not Completely Resolved) | |
---|---|
Possible Cause | Remedy |
Insufficient run time |
|
Incorrect gel pore size |
|
Leftmost and/or Rightmost Bands of the Gel are Distorted | |
---|---|
Possible Cause | Remedy |
Edge effects |
|
Bands Form Smiling Shapes | |
---|---|
Possible Cause | Remedy |
Excessive heat ![]() Gel run is “smiling.” This is caused by too high a voltage. Also, target bands are bleached, caused by using the detection reagent at too high of a concentration. |
|
Network error: Failed to fetch
Network error: Failed to fetch