Merck
LC/MS/MS Analysis of Pesticide Residues in Dates on the Ascentis® Express RP-Amide Column after QuEChERS Extraction
LC/MS/MS Analysis of Pesticide Residues in Dates on the Ascentis® Express RP-Amide Column after QuEChERS Extraction application for LC-MS, application for SPE
Materials
analytical column
Product No.
Description
Pricing
Ascentis® Express RP-Amide, 2 μm UHPLC Column
2 μm particle size, L × I.D. 10 cm × 2.1 mm
accessory
Product No.
Description
Pricing
Certified Vial Kit, Low Adsorption (LA), 2 mL, pk of 100
volume 2 mL, amber glass vial (with marking spot), natural PTFE/silicone septa (with slit), thread for 9 mm
Benchmixer XL multi-tube vortexer
AC/DC input 115 V AC
sample prep
Product No.
Description
Pricing
Supel QuE
Citrate Extraction Tube, pk of 50, suitable for EN 15662
Supel QuE
Empty Centrifuge Tube with Lid, centrifuge tube volume 50 mL, pk of 50, suitable for EN 15662:2008 per BS
Supel QuE
PSA/C18 Tube, pack of 100, AOAC® 2007.01
Supel QuE
PSA Tube, pk of 50, AOAC® 2007.01, centrifuge tube volume 15 mL , Shaker Compatible
standard
Product No.
Description
Pricing
Quinalphos
PESTANAL®, analytical standard
Rotenone
PESTANAL®, analytical standard
Aldicarb-sulfone
PESTANAL®, analytical standard
Etrimfos
PESTANAL®, analytical standard
Neburon
PESTANAL®, analytical standard
Omethoate
PESTANAL®, analytical standard
Butocarboximsulfoxide
PESTANAL®, analytical standard
Ethylene oxide solution
certified reference material, 50 mg/mL in methanol
Spiromesifen
PESTANAL®, analytical standard
Malathion
certified reference material, TraceCERT®
Carbendazim
certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Aldicarb
certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Methomyl
certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Thiacloprid
certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Carbofuran
reference material
Bifenazate
reference material
Chlorantraniliprole
reference material
Methabenzthiazuron
certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Triazophos
certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Trichlorfon
certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Spinetoram solution
100 μg/mL in acetonitrile, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Pyraflufen-ethyl
certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
substrate
Product No.
Description
Pricing
Glufosinate-ammonium
PESTANAL®, analytical standard
CONDITIONS
sample/matrix
5 g ground dates spiked with pesticides at 10 ng/g
extraction process
add 10 mL water and 10 mL acetonitrile; shake 1 hr at 2500 rpm; add contents of Supel QuE citrate extraction tube (55227-U); . (shake for 1 minute; centrifuge at 5000 rpm fo 5 minutes and draw off supernatant)
clean-up tube
Supel QuE PSA/C18 (55288-U)
clean-up process
transfer 1 mL of the extract to 2 mLSupel QuE PSA/C18 tube; shake for 1 minute; centrifuge at 5000 rpm for 3 min; draw off supernatant for analysis.
sample preparation
Dilute 100 μL of cleaned extract to a final volume of 1 mL in 5 mM ammonium formate/0.1% formic acid in water; transfer diluted sample to vial for LC/MS/MS analysis
column
Ascentis Express RP-Amide, 10 cm x 2.1 mm I.D., 2 μm particles (51576-U)
mobile phase
(A) 5 mM ammonium formate with 0.1% formic acid in water; (B) 5 mM ammonium formate with 0.1% formic acid in 95:5 acetonitrile:H2O
gradient
held at 5% B from 0 to 1 min; to 100% B in 13 min; held at 100% B for 1.5 min; to 95% A in 0.5 min; held at 95% A for 1.5 min
flow rate
0.4 mL/min
column temp.
30 °C
detector
MS, ESI (+), MRM
injection
5 μL
Description
Application
Freeze dates with liquid nitrogen and grind to a homogenous mixture.
Analysis Note
The “quick, easy, cheap, effective, rugged, and safe” (QuEChERS) method can be used to analyze multiple pesticide residues found in dates. In this study, QuEChERS extraction and cleanup using PSA/C18 were used for the analysis of pesticide residues of interest in pistachios. Average recoveries obtained for replicate samples spiked at 10 ng/g were in the range of 70-120% for 23 of the 25 pesticides analyzed.
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany