US EPA Method TO-17: GC Analysis of Volatiles on Equity-1 after Collection/Desorption using Carbotrap 300 Tube
CONDITIONS
thermal desorption tube
Carbotrap® 300 glass TD tube, fritted, O.D. × L 1/4 in. × 3 1/2 in., unconditioned, pkg of 10 ea (30141-U)
sample/matrix
TO15/17 Calibration Mix, 1 PPM in N2 (41973-U)
dry purge
1 min
bake
350 °C
transfer line/valve temp.
175 °C
column
FS CAP Equity-1 30 m x 0.25 mm I.D. x 1.0 μm (28049-U)
oven
40 °C/8 min to 100 °C @ 5 °C/min to 220 °C @ 15 °C/min, hold for 10 min
detector
MSD
MSD interface
175 °C
scan range
35-265
carrier gas
Helium
Description
Analysis Note
This application shows the chromatography achieved using an Equity-1 column.
Legal Information
Carbotrap is a registered trademark of Merck KGaA, Darmstadt, Germany
null