Merck
  • Separation of pairs of C-24 epimeric sterols by glass capillary gas liquid chromatography.
Separation of pairs of C-24 epimeric sterols by glass capillary gas liquid chromatography.
Lipids (1981-09-01)
R H Thompson, G Patterson, M J Thompson, H T Slover
ABSTRACT
Paris of C-24 epimeric sterols have been very difficult to separate by physical emthods. We report here the partial or complete separation of the trimethylsilyl ethers of nine pairs of C-24 epimeric sterols by gas liquid chromatography on a glass capillary column coated with SP-2340. The trimethylsilyl ethers of the epimeric pairs of sterols with saturated side chains and a pair with two double bonds in the side chain were completely separated from each other by GLC. The epimeric pairs with a double bond at C-22 showed partial separation. The 24β-epimer with a saturated side chain eluted before the corresponding 24α-epimer. This order was reversed for pairs of C-24 epimeric sterol trimethylsilyl ethers containing a double bond in the side chain at C-22.
Product Description
Supelco
SP®-2340 Capillary GC Column, L × I.D. 30 m × 0.25 mm, df 0.20 μm
Supelco
SP®-2340 Capillary GC Column, L × I.D. 60 m × 0.25 mm, df 0.20 μm
Supelco
SP®-2340 Capillary GC Column, L × I.D. 60 m × 0.32 mm, df 0.20 μm