Merck
  • Lewis Acid Induced Toggle from Ir(II) to Ir(IV) Pathways in Photocatalytic Reactions: Synthesis of Thiomorpholines and Thiazepanes from Aldehydes and SLAP Reagents.
Lewis Acid Induced Toggle from Ir(II) to Ir(IV) Pathways in Photocatalytic Reactions: Synthesis of Thiomorpholines and Thiazepanes from Aldehydes and SLAP Reagents.
ACS central science (2017-02-06)
Sheng-Ying Hsieh, Jeffrey W Bode
ABSTRACT
Redox neutral photocatalytic transformations often require careful pairing of the substrates and photoredox catalysts in order to achieve a catalytic cycle. This can limit the range of viable transformations, as we recently observed in attempting to extend the scope of the photocatalytic synthesis of N-heterocycles using silicon amine protocol (SLAP) reagents to include starting materials that require higher oxidation potentials. We now report that the inclusion of Lewis acids in photocatalytic reactions of organosilanes allows access to a distinct reaction pathway featuring an Ir(III)*/Ir(IV) couple instead of the previously employed Ir(III)*/Ir(II) pathway, enabling the transformation of aromatic and aliphatic aldehydes to thiomorpholines and thiazepanes. The role of the Lewis acid in accepting an electron-either directly or via coordination to an imine-can be extended to other classes of photocatalysts and transformations, including oxidative cyclizations. The combination of light induced reactions and Lewis acids therefore promises access to new pathways and transformations that are not viable using the photocatalysts alone.
Sigma-Aldrich
SLAP N-Bn Pip Reagent, 95%
Sigma-Aldrich
SLAP HydroPyrrolopyrazine Reagent
Sigma-Aldrich
SLAP N-Bn-3Me-Pip Reagent
Sigma-Aldrich
SLAP TA, 95%
Sigma-Aldrich
SLAP 2,3-Bicyclo-(3,4-THF) N-BnPip Reagent, ≥95%