Merck
  • Alkyl Triarylstannanecarbodithioates: Synthesis, Crystal Structures, and Efficiency in RAFT Polymerization.
Alkyl Triarylstannanecarbodithioates: Synthesis, Crystal Structures, and Efficiency in RAFT Polymerization.
Chemistry (Weinheim an der Bergstrasse, Germany) (2017-08-31)
Ihor Kulai, Nathalie Saffon-Merceron, Zoia Voitenko, Stéphane Mazières, Mathias Destarac
ABSTRACT
Eight alkyl triarylstannanecarbodithioates were synthesized starting from the corresponding triarylstannyl chlorides. They were fully characterized by IR and
Sigma-Aldrich
4-Fluorobenzyl bromide, 97%
Sigma-Aldrich
4-Nitrobenzyl bromide, 99%
Sigma-Aldrich
4-Bromotoluene, 98%
Sigma-Aldrich
(1-Bromoethyl)benzene, 97%
Sigma-Aldrich
Methyl 2-bromopropionate, 98%
Sigma-Aldrich
Tin(IV) chloride, 98%