Merck
  • PKC-α Triggers EGFR Ubiquitination, Endocytosis and ERK Activation in Podocytes Stimulated with High Glucose.
PKC-α Triggers EGFR Ubiquitination, Endocytosis and ERK Activation in Podocytes Stimulated with High Glucose.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology (2017-05-24)
Chun-Tao Lei, Yan-Hong Wei, Hui Tang, Qian Wen, Chen Ye, Chun Zhang, Hua Su
ABSTRACT
Protein Kinase C-α (PKC-α) and epidermal growth factor receptor (EGFR) are both involved in diabetic kidney disease; however, the connection between these two proteins during high glucose-induced podocyte injury remains uncertain. Diabetes was induced in SD rats by streptozotocin (STZ). Fourteen days later, the kidney cortex was removed and subjected to plasma membrane isolation and lipid raft fractionation. In vitro study human podocyte cell line was differentiated and subjected to various treatments. The levels of membranous protein and endocytosis were assessed by biotinylation and sodium 2-mercaptoethane sulfonate (MesNa) treatment. Gö6976 and PYR-41 were used as inhibitors of PKC-α and ubiquitin activating E1 enzyme, respectively. In diabetic rats, the abundance of PKC-α in the membranous fraction and the lipid raft domain is elevated, whereas the EGFR level is reduced. Consistently, in vitro high glucose treated podocytes, membranous EGFR is downregulated with increased PKC-α. Furthermore, the ubiquitination and endocytosis of EGFR are enhanced accompanied by extracellular signal-regulated kinase (ERK) signaling activation and podocyte damage during hyperglycemia. However, these processes can be ameliorated by inhibition of either PKC-α or ubiquitin activating E1 enzyme. During hyperglycemia, PKC-α mediates podocytic EGFR ubiquitination, endocytosis from cell surface and the subsequent ERK activation, which contributes to podocyte injury.
Sigma-Aldrich
PYR-41, ≥98% (HPLC), powder
Sigma-Aldrich
Ubiquitin E1 Inhibitor, PYR-41, The Ubiquitin E1 Inhibitor, PYR-41 controls the biological activity of Ubiquitin E1. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.