Merck
  • Equol Attenuates Atherosclerosis in Apolipoprotein E-Deficient Mice by Inhibiting Endoplasmic Reticulum Stress via Activation of Nrf2 in Endothelial Cells.
Equol Attenuates Atherosclerosis in Apolipoprotein E-Deficient Mice by Inhibiting Endoplasmic Reticulum Stress via Activation of Nrf2 in Endothelial Cells.
PloS one (2016-12-03)
Ting Zhang, Qin Hu, Linying Shi, Li Qin, Qianyong Zhang, Mantian Mi
ABSTRACT
The development of atherosclerosis is closely related to excessive endoplasmic reticulum stress (ERs). Equol reportedly protects against cardiovascular disease; however, the underlying mechanism for this protection remains unknown. Herein, the mechanisms contributing to the atheroprotective effect of equol were addressed using apolipoprotein E knockout (apoE-/-) mice fed a high-fat diet (HFD) with or without equol. Equol intervention reduced atherosclerotic lesions in the aorta in HFD-fed apoE-/- mice. Plasma lipid analysis showed that equol intervention reduced triglycerides, total cholesterol and LDL-cholesterol and increased HDL-cholesterol. Additionally, equol administration decreased lipid accumulation in the liver. Simultaneously, equol treatment inhibited cell apoptosis induced by t-BHP and thapsigargin in human umbilical vein endothelial cells (HUVECs). Furthermore, equol treatment attenuated palmitate, t-BHP or thapsigargin-induced upregulation of ER stress markers, including p-PERK, p-eIF2α, GRP78, ATF6 and CHOP proteins expression. The same tendency was also observed in aortic lysates in apoE-/- mice fed with equol plus HFD compared with HFD alone. Moreover, equol treatment dose dependently activated the Nrf2 signaling pathway under oxidative stress. Additionally, elevation of Nrf2 induction was found in aortic lysates in apoE-/- mice fed with a HFD diet containing equol compared with a HFD diet without equol. Importantly, Nrf2 siRNA interference induced CHOP and attenuated the effect of equol to inhibit t-BHP mediated CHOP induction, furthermore, abrogated cell apoptosis induced by t-BHP, suggesting a role for Nrf2 in the protective effect of equol in HUVECs. Collectively, these findings implicate that the improvement of atherosclerosis by equol through attenuation of ER stress is mediated, at least in part, by activating the Nrf2 signaling pathway.
Sigma-Aldrich
Palmitic acid, BioXtra, ≥99%
Sigma-Aldrich
Oil Red O, certified by the Biological Stain Commission
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder
Sigma-Aldrich
Anti-ATF6 (ab2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film