Merck
  • The Electroporation as a Tool for Studying the Role of Plasma Membrane in the Mechanism of Cytotoxicity of Bisphosphonates and Menadione.
The Electroporation as a Tool for Studying the Role of Plasma Membrane in the Mechanism of Cytotoxicity of Bisphosphonates and Menadione.
The Journal of membrane biology (2016-04-06)
Mantas Šilkūnas, Rita Saulė, Danutė Batiuškaitė, Gintautas Saulis
ABSTRACT
In this study, the role of the cell plasma membrane as a barrier in the mechanism of the cytotoxicity of nitrogen-containing bisphosphonates and menadione was studied, and the possibility of increasing the efficiency of bisphosphonates and menadione (vitamin K
Sigma-Aldrich
Gram′s crystal violet solution, for microscopy
Sigma-Aldrich
Trypsin-EDTA solution, 0.25%, sterile-filtered, BioReagent, suitable for cell culture, 2.5 g porcine trypsin and 0.2 g EDTA, 4Na per liter of Hanks′ Balanced Salt Solution with phenol red