Merck
  • Bioactive nanofibers enable the identification of thrombospondin 2 as a key player in enamel regeneration.
Bioactive nanofibers enable the identification of thrombospondin 2 as a key player in enamel regeneration.
Biomaterials (2015-05-26)
Zhan Huang, Christina J Newcomb, Yaping Lei, Yan Zhou, Paul Bornstein, Brad A Amendt, Samuel I Stupp, Malcolm L Snead
ABSTRACT
Tissue regeneration and development involves highly synchronized signals both between cells and with the extracellular environment. Biomaterials can be tuned to mimic specific biological signals and control cell response(s). As a result, these materials can be used as tools to elucidate cell signaling pathways and candidate molecules involved with cellular processes. In this work, we explore enamel-forming cells, ameloblasts, which have a limited regenerative capacity. By exposing undifferentiated cells to a self-assembling matrix bearing RGDS epitopes, we elicited a regenerative signal at will that subsequently led to the identification of thrombospondin 2 (TSP2), an extracellular matrix protein that has not been previously recognized as a key player in enamel development and regeneration. Targeted disruption of the thrombospondin 2 gene (Thbs2) resulted in enamel formation with a disordered architecture that was highly susceptible to wear compared to their wild-type counterparts. To test the regenerative capacity, we injected the bioactive matrix into the enamel organ and discovered that the enamel organic epithelial cells in TSP-null mice failed to polarize on the surface of the artificial matrix, greatly reducing integrin β1 and Notch1 expression levels, which represent signaling pathways known to be associated with TSP2. These results suggest TSP2 plays an important role in regulating cell-matrix interactions during enamel formation. Exploiting the signaling pathways activated by biomaterials can provide insight into native signaling mechanisms crucial for tooth development and cell-based strategies for enamel regeneration.
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
Sodium orthovanadate, 99.98% trace metals basis
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
3-Amino-9-ethylcarbazole, ≥95% (HPLC), powder
Sigma-Aldrich
3-Amino-9-ethylcarbazole, tablet
Sigma-Aldrich
Thrombopoietin from mouse, recombinant, expressed in NSO cells, lyophilized powder, suitable for cell culture, >97% (SDS-PAGE)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
Sodium orthovanadate, ≥90% (titration)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0%