Merck
  • Effect of starch-beeswax coatings on quality parameters of blackberries (Rubus spp.).
Effect of starch-beeswax coatings on quality parameters of blackberries (Rubus spp.).
Journal of food science and technology (2015-09-08)
Alfonso Pérez-Gallardo, Blanca García-Almendárez, Gustavo Barbosa-Cánovas, Diana Pimentel-González, L R Reyes-González, Carlos Regalado
ABSTRACT
There is increased interest in berry fruits due to health benefits, and maintenance of fruit quality for longer periods of time has been a priority. We previously found that starch based coatings applied on raspberries was associated to volatile compounds production due to anoxic conditions. The objective of this work was to design more hydrophobic coatings with reduced thickness. A starch-beeswax dispersion containing 2 % (w/v) modified tapioca starch added with either 0.5 or 1.0 % (w/v) beeswax microparticles was produced, and used for spray coating freshly harvested blackberries (Rubus spp.). Coatings were air dried, packed in plastic trays and stored up to 16 days at 4 °C and 88 % relative humidity. Storage quality parameters such as hardness, respiration rate, anthocyanins content, total phenols, color changes and weight loss were evaluated. We did not find Interactions among coating ingredients, and incorporation of beeswax reduced moisture transfer rate. Coatings did not occlude the stomata and apparently did not over-hydrate the cuticle. This characteristic allowed appropriate gas exchange (O2 and CO2), and reduced accumulation of volatile compounds associated to fermentative metabolism. Respiration rates were 4.207 ± 0.157, 4.557 ± 0.220 and 4.780 ± 0.050 mmol CO2 kg(-1) h(-1) for control, 0.5 and 1 % of wax content in coatings, respectively. However, ethylene production increased throughout storage time along with beeswax concentration, indicating stressful conditions for the fruit. This trend appears to be related with changes in total phenols and anthocyanins during storage. Edible coatings based on starch and hydrophobic particles should be reformulated to maintain quality of stored berry fruits.
Sigma-Aldrich
Ethylene, 99.99%
Sigma-Aldrich
Morpholine, purified by redistillation, ≥99.5%
Sigma-Aldrich
D-Sorbitol, FCC, FG
Sigma-Aldrich
D-Sorbitol, 99% (GC)
Sigma-Aldrich
Ethylene, ≥99.5%
Sigma-Aldrich
Morpholine, ReagentPlus®, ≥99%
Sigma-Aldrich
D-Sorbitol, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
Sorbitol F solution, 70 wt. % in H2O, Contains mainly D-sorbitol with lesser amounts of other hydrogenated oligosaccharides
Sigma-Aldrich
Morpholine, ACS reagent, ≥99.0%
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioXtra
Sigma-Aldrich
D-Sorbitol, ≥98% (GC)
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), for molecular biology
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Stearic acid, SAJ first grade, ≥90.0%, powder
Sigma-Aldrich
Stearic acid, reagent grade, 95%
Sigma-Aldrich
Stearic acid, Grade I, ≥98.5% (capillary GC)
Sigma-Aldrich
Stearic acid, ≥95%, FCC, FG
Sigma-Aldrich
StratoSpheres PL-MPH resin, 50-100 mesh, extent of labeling: 3.0-4.0 mmol/g loading, 1 % cross-linked
Sigma-Aldrich
Morpholine, polymer-bound, 200-400 mesh, extent of labeling: 2.75-3.25 mmol/g loading, 1 % cross-linked