Merck
  • The development of dimple-shaped chitosan carrier for ethambutol dihydrochloride dry powder inhaler.
The development of dimple-shaped chitosan carrier for ethambutol dihydrochloride dry powder inhaler.
Drug development and industrial pharmacy (2014-04-04)
Md Iftekhar Ahmad, Suwipa Ungphaiboon, Teerapol Srichana
ABSTRACT
In this study, a dimple-shaped chitosan carrier was developed for delivering the antituberculosis drug ethambutol dihydrochloride (EDH) from a dry powder inhaler (DPI) to the lungs. Nanosized drug particles were prepared using nanospray drying. The microsized carrier was developed from a chitosan solution by spray drying. Five formulations were prepared by physically mixing the drug and carrier in different ratios. The physico-chemical properties of the formulations were analyzed using scanning electron microscopy, differential scanning calorimetry, Fourier transform infrared spectroscopy, ultracentrifugation and a cascade impactor. The EDH size was 222 nm and the chitosan carrier size was 1.2 µm. Five formulations, i.e. 1:2, 1:2.5, 1:3.3, 1:5 and 1:10 w/w of the EDH to chitosan carrier were prepared by physical mixing. The chitosan carrier was spherical in shape with a dimpled surface and this provided shallow cavities to which the drug was bound, both within its grooves as well as on its surface. The median adhesion force (50% of drug detachment) for formulations #1 to #5 was between 122 and 993 µN. The mass median aerodynamic diameter of the EDH was between 2.3 and 2.7 μm, with the fine particle fractions (aerosolized particles less than 4.4 μm) of 32-42% of the nominal dose. We suggest that ethambutol dihydrochloride mixed with a chitosan carrier was suitable for use in a dry powder inhaler for controlling tuberculosis especially in minimizing the risk of multidrug resistant tuberculosis and the possible side effects from EDH.
Sigma-Aldrich
Acetic acid, 99.5-100.0%
Sigma-Aldrich
Acetic acid, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetic acid, JIS special grade, ≥99.7%
Sigma-Aldrich
Acetic acid, ≥99.7%
Sigma-Aldrich
Acetic acid, ≥99.7%, suitable for amino acid analysis
Sigma-Aldrich
Acetic acid, ≥99.7%
Sigma-Aldrich
Acetic acid solution, 1 M, 1 N
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Sodium 1-heptanesulfonate monohydrate, ≥96.0% (T)
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%